Merck
Solvents in water at 20p
Solvents in water at 20p suitable for GC, application for SPME
Materiales
Fibra para SPME
Referencia del producto
Descripción
Precios
Fibra de carboxeno/polidimetilsiloxano (DVB/CAR/PDMS) de SPME
df 75 μm(CAR/PDMS, for use with manual holder, needle size 24 ga
columna analítica
Referencia del producto
Descripción
Precios
SPB®-1 Sulfur
L × I.D. 30 m × 0.32 mm, df 4.00 μm
CONDITIONS
sample/matrix
solvents at 20 ppb in 4 mL water + 25% NaCl in 4 mL vial
SPME fiber
carboxen/PDMS, 75 μm (57318)
extraction
immersion, 10 min, rapid stirring
desorption process
270 °C, 5 min
column
SPB-1 SULFUR, 30 m × 0.32 mm I.D., 4.0 μm film (24158)
oven
50°C (2 min) to 150°C at 10°C/min
inj. temp.
splitless (closed 2 min), 260°C, 0.75 mm I.D. liner
detector
FID
carrier gas
helium, 30 cm/sec
Descripción
Información legal
SPB is a registered trademark of Merck KGaA, Darmstadt, Germany