Merck
PP2A Inactivation Mediated by
Cancer research (2017-10-20)
Ward Sents, Bob Meeusen, Petar Kalev, Enrico Radaelli, Xavier Sagaert, Eline Miermans, Dorien Haesen, Caroline Lambrecht, Mieke Dewerchin, Peter Carmeliet, Jukka Westermarck, Anna Sablina, Veerle Janssens
ABSTRACT
Protein phosphatase 2A (PP2A) complexes counteract many oncogenic kinase pathways. In cancer cells, PP2A function can be compromised by several mechanisms, including sporadic mutations in its scaffolding A and regulatory B subunits or more frequently through overexpression of cellular PP2A inhibitors. Here, we identify a novel genetic mechanism by which PP2A function is recurrently affected in human cancer, involving haploinsufficiency of