Merck
  • Development of a radiolabeled caninized anti-EGFR antibody for comparative oncology trials.
Development of a radiolabeled caninized anti-EGFR antibody for comparative oncology trials.
Oncotarget (2017-11-16)
Judit Fazekas-Singer, Neydher Berroterán-Infante, Christina Rami-Mark, Monika Dumanic, Miroslawa Matz, Michael Willmann, Fritz Andreae, Josef Singer, Wolfgang Wadsak, Markus Mitterhauser, Erika Jensen-Jarolim
ABSTRACT
Due to large homology of human and canine EGFR, dogs suffering from spontaneous EGFR+ cancer can be considered as ideal translational models. Thereby, novel immunotherapeutic compounds can be developed for both human and veterinary patients. This study describes the radiolabeling of a canine anti-EGFR IgG antibody (can225IgG) with potential diagnostic and therapeutic value in comparative clinical settings. Can225IgG was functionalized with DTPA for subsequent chelation with the radionuclide
Sigma-Aldrich
Fluoromount Aqueous Mounting Medium, for use with fluorescent dye-stained tissues
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline