Merck
  • The development of a novel immunotherapy model of human ovarian cancer in human PBL-severe combined immunodeficient (SCID) mice.
The development of a novel immunotherapy model of human ovarian cancer in human PBL-severe combined immunodeficient (SCID) mice.
Clinical and experimental immunology (1995-09-01)
W Walker, G Gallagher
ABSTRACT
The reported ability of SCID mice to accept xenografts of both human tumors and peripheral blood lymphocytes (PBL) provides the potential for the development of novel immunotherapy models in these animals. This study describes the development of a novel small animal model of human ovarian cancer. This was achieved by engrafting a human ovarian cancer cell line (Ovan-4) into the peritoneal cavity of immunodeficient SCID and immune reconstituted human PBL-SCID mice. When transplanted to SCID mice this cell line exhibited growth characteristics similar to the clinical disease observed in patients with implantation of metastatic nodules onto the interior surface of the peritoneal wall. Reconstituted human PBL-SCID mice challenged with identical numbers of Ovan-4 cells exhibited a significant increase in survival time, suggesting a role for cells of the human immune system in preventing the development of this type of malignancy in vivo. Furthermore, vaccination of human PBL-SCID mice against Ovan-4 produced tumour-specific human antibodies in the serum of these animals. Animals reconstituted with CD8-depleted PBL exhibited increased serum immunoglobulin levels and produced enhanced anti-Ovan-4 activity after vaccination. Subsequent challenge of these animals with Ovan-4 revealed a further increase in survival time. These results suggest that human antibodies may have a role in immunity against ovarian cancer and could be of therapeutic value in this type of disease.