Merck
  • The defect of SFRP2 modulates an influx of extracellular calcium in B lymphocytes.
The defect of SFRP2 modulates an influx of extracellular calcium in B lymphocytes.
BMC research notes (2014-11-06)
Yuichi Tokuda, Masami Tanaka, Tomohito Yagi, Kei Tashiro
ABSTRACT
In the Wnt pathway, the secreted frizzled-related protein 2 (SFRP2) is thought to act as one of the several competitive inhibitors of Wnt. However, the precise role of SFRP2 is still poorly understood especially in B lymphocytes. Here, we investigated the function of SFRP2, comparing the SFRP2 defective as well as normal B lymphocytes in mice. We demonstrated that calcium influx from extracellular to intracellular space in splenic B cells was clearly affected by the defect of SFRP2. In addition, the phosphorylation of phospholipase Cγ2 was observed to be reduced in SFRP2 defective splenic B cells with B cell receptor stimulation. SFRP2 is suggested to modulate the influx from extracellular calcium in the B cell receptor signaling pathway.