Merck
  • Identification of proteomic biomarkers predicting prostate cancer aggressiveness and lethality despite biopsy-sampling error.
Identification of proteomic biomarkers predicting prostate cancer aggressiveness and lethality despite biopsy-sampling error.
British journal of cancer (2014-07-18)
M Shipitsin, C Small, S Choudhury, E Giladi, S Friedlander, J Nardone, S Hussain, A D Hurley, C Ernst, Y E Huang, H Chang, T P Nifong, D L Rimm, J Dunyak, M Loda, D M Berman, P Blume-Jensen
ABSTRACT
Key challenges of biopsy-based determination of prostate cancer aggressiveness include tumour heterogeneity, biopsy-sampling error, and variations in biopsy interpretation. The resulting uncertainty in risk assessment leads to significant overtreatment, with associated costs and morbidity. We developed a performance-based strategy to identify protein biomarkers predictive of prostate cancer aggressiveness and lethality regardless of biopsy-sampling variation. Prostatectomy samples from a large patient cohort with long follow-up were blindly assessed by expert pathologists who identified the tissue regions with the highest and lowest Gleason grade from each patient. To simulate biopsy-sampling error, a core from a high- and a low-Gleason area from each patient sample was used to generate a 'high' and a 'low' tumour microarray, respectively. Using a quantitative proteomics approach, we identified from 160 candidates 12 biomarkers that predicted prostate cancer aggressiveness (surgical Gleason and TNM stage) and lethal outcome robustly in both high- and low-Gleason areas. Conversely, a previously reported lethal outcome-predictive marker signature for prostatectomy tissue was unable to perform under circumstances of maximal sampling error. Our results have important implications for cancer biomarker discovery in general and development of a sampling error-resistant clinical biopsy test for prediction of prostate cancer aggressiveness.