Merck
  • High-resolution, noninvasive longitudinal live imaging of immune responses.
High-resolution, noninvasive longitudinal live imaging of immune responses.
Proceedings of the National Academy of Sciences of the United States of America (2011-07-20)
Midhat H Abdulreda, Gaetano Faleo, Ruth Damaris Molano, Maite Lopez-Cabezas, Judith Molina, Yaohong Tan, Oscar A Ron Echeverria, Elsie Zahr-Akrawi, Rayner Rodriguez-Diaz, Patrick K Edlund, Ingo Leibiger, Allison L Bayer, Victor Perez, Camillo Ricordi, Alejandro Caicedo, Antonello Pileggi, Per-Olof Berggren
ABSTRACT
Intravital imaging emerged as an indispensible tool in biological research, and a variety of imaging techniques have been developed to noninvasively monitor tissues in vivo. However, most of the current techniques lack the resolution to study events at the single-cell level. Although intravital multiphoton microscopy has addressed this limitation, the need for repeated noninvasive access to the same tissue in longitudinal in vivo studies remains largely unmet. We now report on a previously unexplored approach to study immune responses after transplantation of pancreatic islets into the anterior chamber of the mouse eye. This approach enabled (i) longitudinal, noninvasive imaging of transplanted tissues in vivo; (ii) in vivo cytolabeling to assess cellular phenotype and viability in situ; (iii) local intervention by topical application or intraocular injection; and (iv) real-time tracking of infiltrating immune cells in the target tissue.
Sigma-Aldrich
TAK-779, ≥98% (HPLC)