Merck
  • Role of the 3(ππ*) state in photolysis of lumisantonin: insight from ab initio studies.
Role of the 3(ππ*) state in photolysis of lumisantonin: insight from ab initio studies.
The journal of physical chemistry. A (2011-06-02)
Xing Chen, Zilvinas Rinkevicius, Yi Luo, Hans Ågren, Zexing Cao
ABSTRACT
The CASSCF and CASPT2 methodologies have been used to explore the potential energy surfaces of lumisantonin in the ground and low-lying triplet states along the photoisomerization pathways. Calculations indicate that the (1)(nπ*) state is the accessible low-lying singlet state with a notable oscillator strength under an excitation wavelength of 320 nm and that it can effectively decay to the (3)(ππ*) state through intersystem crossing in the region of minimum surface crossings with a notable spin-orbital coupling constant. The (3)(ππ*) state, derived from the promotion of an electron from the π-type orbital mixed with the σ orbital localized on the C-C bond in the three-membered alkyl ring to the π* orbital of conjugation carbon atoms, plays a critical role in C-C bond cleavage. Based on the different C-C bond rupture patterns, the reaction pathways can be divided into paths A and B. Photolysis along path A arising from C1-C5 bond rupture is favorable because of the dynamic and thermodynamic preferences on the triplet excited-state PES. Path B is derived from the cleavage of the C5-C6 bond, leading first to a relatively stable species, compared to intermediate A-INT formed on the ground state PES. Accordingly, path B is relatively facile for the pyrolytic reaction. The present results provide a basis to interpret the experimental observations.