Merck
  • Pooled CRISPR screening identifies m6A as a positive regulator of macrophage activation.
Pooled CRISPR screening identifies m6A as a positive regulator of macrophage activation.
Science advances (2021-04-30)
Jiyu Tong, Xuefei Wang, Yongbo Liu, Xingxing Ren, Anmin Wang, Zonggui Chen, Jiameng Yao, Kaiqiong Mao, Tingting Liu, Fei-Long Meng, Wen Pan, Qiang Zou, Jun Liu, Yu Zhou, Qiang Xia, Richard A Flavell, Shu Zhu, Hua-Bing Li
ABSTRACT
m6A RNA modification is implicated in multiple cellular responses. However, its function in the innate immune cells is poorly understood. Here, we identified major m6A "writers" as the top candidate genes regulating macrophage activation by LPS in an RNA binding protein focused CRISPR screening. We have confirmed that Mettl3-deficient macrophages exhibited reduced TNF-α production upon LPS stimulation in vitro. Consistently, Mettl3flox/flox;Lyzm-Cre mice displayed increased susceptibility to bacterial infection and showed faster tumor growth. Mechanistically, the transcripts of the Irakm gene encoding a negative regulator of TLR4 signaling were highly decorated by m6A modification. METTL3 deficiency led to the loss of m6A modification on Irakm mRNA and slowed down its degradation, resulting in a higher level of IRAKM, which ultimately suppressed TLR signaling-mediated macrophage activation. Our findings demonstrate a previously unknown role for METTL3-mediated m6A modification in innate immune responses and implicate the m6A machinery as a potential cancer immunotherapy target.