- Interleukin-34 Levels Were Associated with Prognosis in Patients with Acute Myocardial Infarction.
Interleukin-34 Levels Were Associated with Prognosis in Patients with Acute Myocardial Infarction.
Inflammatory factors have specific value in acute myocardial infarction (AMI). Our previous studies have identified the prognostic value of interleukin (IL)-34 during chronic heart failure. However, the potential impact of IL-34 on AMI remains unknown.Serum IL-34 was measured in 287 AMI patients, and they were followed up for the composite endpoint, including cardiovascular death, heart failure hospitalization, recurrent nonfatal myocardial infarction (MI), and nonfatal stroke.IL-34 levels were significantly associated with the presence of heart failure at baseline and its aggravation after a year. During the five-year follow-up, there was a significant increase in the risk of the composite endpoint (hazard ratio [HR] 1.38 [95% confidence intervals (CI) 1.12-1.70], P < 0.01) and cardiovascular death (HR 1.48 [95%CI 1.03-2.27], P = 0.03) after full adjustment as IL-34 levels increased.Higher IL-34 levels in the acute phase were associated with an increased risk of heart failure after MI and poor prognosis.