Merck
  • Ultrafast Two-Photon Imaging of a High-Gain Voltage Indicator in Awake Behaving Mice.
Ultrafast Two-Photon Imaging of a High-Gain Voltage Indicator in Awake Behaving Mice.
Cell (2019-12-14)
Vincent Villette, Mariya Chavarha, Ivan K Dimov, Jonathan Bradley, Lagnajeet Pradhan, Benjamin Mathieu, Stephen W Evans, Simon Chamberland, Dongqing Shi, Renzhi Yang, Benjamin B Kim, Annick Ayon, Abdelali Jalil, François St-Pierre, Mark J Schnitzer, Guoqiang Bi, Katalin Toth, Jun Ding, Stéphane Dieudonné, Michael Z Lin
ABSTRACT
Optical interrogation of voltage in deep brain locations with cellular resolution would be immensely useful for understanding how neuronal circuits process information. Here, we report ASAP3, a genetically encoded voltage indicator with 51% fluorescence modulation by physiological voltages, submillisecond activation kinetics, and full responsivity under two-photon excitation. We also introduce an ultrafast local volume excitation (ULoVE) method for kilohertz-rate two-photon sampling in vivo with increased stability and sensitivity. Combining a soma-targeted ASAP3 variant and ULoVE, we show single-trial tracking of spikes and subthreshold events for minutes in deep locations, with subcellular resolution and with repeated sampling over days. In the visual cortex, we use soma-targeted ASAP3 to illustrate cell-type-dependent subthreshold modulation by locomotion. Thus, ASAP3 and ULoVE enable high-speed optical recording of electrical activity in genetically defined neurons at deep locations during awake behavior.
Sigma-Aldrich
CNQX disodium salt hydrate, ≥98% (HPLC), solid