Merck
  • Allosensitization after transplant failure: the role of graft nephrectomy and immunosuppression - a retrospective study.
Allosensitization after transplant failure: the role of graft nephrectomy and immunosuppression - a retrospective study.
Transplant international : official journal of the European Society for Organ Transplantation (2019-04-14)
Gaetano Lucisano, Paul Brookes, Eva Santos-Nunez, Nicola Firmin, Nicola Gunby, Sevda Hassan, Alexander Gueret-Wardle, Paul Herbert, Vassilios Papalois, Michelle Willicombe, David Taube
ABSTRACT
There are conflicting data about the role of transplant nephrectomy and immunosuppression withdrawal on the development of allosensitization and the impact on re-transplantation. We divided 109 first graft recipients into two groups according to whether they underwent nephrectomy (NX+, n = 61) or their graft was left in situ (NX-, n = 48). Sera were assessed for HLA-A/B/Cw/DR/DQ antibodies at the time of NX/transplant failure and after 3, 6, 12, 24 months. The NX+ group showed a higher rate of donor specific antibody (DSA) and non-DSA human leukocyte antigen (HLA) antibody production at all the time points. Multivariable analysis showed that nephrectomy was a strong, independent risk factor for the development of DSAs after 12 and 24 months (P = 0.005 and 0.008). In the NX- group, low tacrolimus levels correlated with DSA formation (AUC 0.817, P = 0.002; best cut-off level 2.9 ng/ml). Analysis with a standardized pool of UK donors showed a more difficult grade of HLA matchability following nephrectomy compared with the NX- group. Nephrectomy is followed by the long-term production of DSA and non-DSA HLA antibodies and negatively impacts on the chances of finding a HLA-compatible kidney. Tacrolimus levels ≥3 ng/ml are protective against the development of allosensitization and could facilitate re-transplantation in the NX- group.