- Lactulose synergizes with CpG-ODN to modulate epithelial and immune cells cross talk.
Lactulose synergizes with CpG-ODN to modulate epithelial and immune cells cross talk.
Food & function (2019-01-12)
R Mukherjee, M van de Kaa, J Garssen, R J Pieters, A D Kraneveld, L E M Willemsen
PMID30632580
ABSTRACT
Lactulose, a non-digestible oligosaccharide and functional food, promotes Bifidobacteria growth. Here we show that lactulose, beyond its prebiotic action, may have direct immunomodulatory effects as well. In synergy with CpG-ODN, a bacterial DNA mimetic, lactulose enhances basolateral concentrations of IFN-γ, IL-10, and galectin-9 in the co-culture model of epithelial and immune cells.