Merck
  • Semaphorin 3C regulates endothelial cell function by increasing integrin activity.
Semaphorin 3C regulates endothelial cell function by increasing integrin activity.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2006-08-31)
Nazifa Banu, Jason Teichman, Marya Dunlap-Brown, Guillermo Villegas, Alda Tufro
ABSTRACT
Class 3 semaphorins (sema 3) are secreted guidance proteins. Sema 3A expressed by endothelial cells controls vascular morphogenesis through integrin inhibition. Sema 3C is required for normal cardiovascular patterning. Here we examined the potential role of sema 3C as regulator of endothelial cell function in vitro using mouse glomerular endothelial cells (MGEC). We determined that MGEC express sema 3C mRNA and protein and its receptors mRNA. Recombinant sema 3C induced MGEC proliferation 18 +/- 2% above control, as assessed by bromodeoxyuridine (BrdU) incorporation, and reduced starvation-induced apoptosis by 46 +/- 3%, as indicated by an in situ marker of activated caspase 3. Sema 3C increased MGEC adhesion to fibronectin 79 +/- 13% and to collagen 55 +/- 12% as compared with control. Sema 3C-induced MGEC adhesion was prevented by integrin blocking antibodies and involved beta1 integrin serine phosphorylation. Sema 3C-induced MGEC adhesion and proliferation were similar to those induced by vascular endothelial growth factor (VEGF)-A. Sema 3C induced a 44 +/- 11% increase in MGEC directional migration and stimulated MGEC capillary-like network formation on collagen I gels. Collectively, our data indicate that sema 3C promotes glomerular endothelial cell proliferation, adhesion, directional migration, and tube formation in vitro by stimulating integrin phosphorylation and VEGF120 secretion, functions that are similar to VEGF-A and opposite to sema 3A.
Sigma-Aldrich
Anti-Integrin αVβ3 Antibody, clone LM609, clone LM609, Chemicon®, from mouse