Merck
  • Semaphorin 6D reverse signaling controls macrophage lipid metabolism and anti-inflammatory polarization.
Semaphorin 6D reverse signaling controls macrophage lipid metabolism and anti-inflammatory polarization.
Nature immunology (2018-05-20)
Sujin Kang, Yoshimitsu Nakanishi, Yoshiyuki Kioi, Daisuke Okuzaki, Tetsuya Kimura, Hyota Takamatsu, Shohei Koyama, Satoshi Nojima, Masayuki Nishide, Yoshitomo Hayama, Yuhei Kinehara, Yasuhiro Kato, Takeshi Nakatani, Tomomi Shimogori, Junichi Takagi, Toshihiko Toyofuku, Atsushi Kumanogoh
ABSTRACT
Polarization of macrophages into pro-inflammatory or anti-inflammatory states has distinct metabolic requirements, with mechanistic target of rapamycin (mTOR) kinase signaling playing a critical role. However, it remains unclear how mTOR regulates metabolic status to promote polarization of these cells. Here we show that an mTOR-Semaphorin 6D (Sema6D)-Peroxisome proliferator receptor γ (PPARγ) axis plays critical roles in macrophage polarization. Inhibition of mTOR or loss of Sema6D blocked anti-inflammatory macrophage polarization, concomitant with severe impairments in PPARγ expression, uptake of fatty acids, and lipid metabolic reprogramming. Macrophage expression of the receptor Plexin-A4 is responsible for Sema6D-mediated anti-inflammatory polarization. We found that a tyrosine kinase, c-Abl, which associates with the cytoplasmic region of Sema6D, is required for PPARγ expression. Furthermore, Sema6D is important for generation of intestinal resident CX3CR1hi macrophages and prevents development of colitis. Collectively, these findings highlight crucial roles for Sema6D reverse signaling in macrophage polarization, coupling immunity, and metabolism via PPARγ.
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Plxna4
Sigma-Aldrich
Dasatinib