![TSKgel® G5000PW HPLC Column phase hydroxylated methacrylate, L × I.D. 60 cm × 7.5 mm, 17 μm particle size](/deepweb/assets/sigmaaldrich/product/images/288/015/a81c4c54-c7a3-4e84-860f-42111d69db53/640/a81c4c54-c7a3-4e84-860f-42111d69db53.jpg)
TSKgel® G5000PW (17 µm) HPLC Columns
![TSKgel® G5000PW HPLC Column phase hydroxylated methacrylate, L × I.D. 60 cm × 7.5 mm, 17 μm particle size](/deepweb/assets/sigmaaldrich/product/images/288/015/a81c4c54-c7a3-4e84-860f-42111d69db53/640/a81c4c54-c7a3-4e84-860f-42111d69db53.jpg)
Synonym(s):
TSKgel® Size Exclusion (PW-Type) HPLC Column
Products
Product Category
Manufacturer
Product Line
Agency
USP Suitability
Separation Technique
ID (mm)
Length (cm)
Particle Size (µm)
Pore Size (Å)
Technique
Showing 1-2 of 2