Skip to Content
Merck
vibrant-m

SUPELCOGEL C610H, 6% Crosslinked HPLC Column

vibrant-m
Synonym(s):
SUPELCOGEL 6% Cross Linked HPLC Columns

Products

Product Category
Product Line
Agency
USP Suitability
Separation Technique
ID (mm)
Length (cm)
Quality Segment
Column Material
Package Size
Particle Size (µm)
Technique
Showing 1-1 of 1