Skip to Content
Merck
K-411 Glass microspheres NIST® SRM® 2066

K-411 Glass microspheres

K-411 Glass microspheres NIST® SRM® 2066
Synonym(s):
Glass spheres
Empirical Formula (Hill Notation):
O2Si
CAS Number:
Molecular Weight:
60.08

Products

Formula Weight
Manufacturer
Physical Form
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique