Skip to Content
Merck
Cetyl palmitate 15 European Pharmacopoeia (EP) Reference Standard

Cetyl Palmitate

Cetyl palmitate 15 European Pharmacopoeia (EP) Reference Standard
Synonym(s):
Palmityl palmitate, Cetyl palmitate, Hexadecyl hexadecanoate, Palmitic acid palmityl ester
Linear Formula:
CH3(CH2)14CO2(CH2)15CH3
CAS Number:
Molecular Weight:
480.85

Products

Brand
Formula Weight
Manufacturer
Melting Point (°C)
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique