Skip to Content
Merck
Phytomenadione British Pharmacopoeia (BP) Reference Standard

Phylloquinone (K1)

Phytomenadione British Pharmacopoeia (BP) Reference Standard
Synonym(s):
Vitamin K1, 2-Methyl-3-phytyl-1,4-naphthoquinone, 3-Phytylmenadione, Phylloquinone, Vitamin K1(20)
Empirical Formula (Hill Notation):
C31H46O2
CAS Number:
Molecular Weight:
450.70
EC Number:

Products

Product Category
Brand
Biological Source
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique