Skip to Content
Merck
Chlorphenamine maleate European Pharmacopoeia (EP) Reference Standard

Chlorpheniramine Maleate

Chlorphenamine maleate European Pharmacopoeia (EP) Reference Standard
Synonym(s):
(±)-Chlorpheniramine maleate salt, Chlorpheniramine maleate solution
Empirical Formula (Hill Notation):
C16H19ClN2 · C4H4O4
CAS Number:
Molecular Weight:
390.86

Products

Brand
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique