Methimazole
Synonym(s):
1-Methyl-2-imidazolethiol, 2-Mercapto-1-methylimidazole, Methimazole
Empirical Formula (Hill Notation):
C4H6N2S
CAS Number:
Molecular Weight:
114.17
Products
Brand
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Product Line
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Special Grade
Technique
Showing 1-5 of 5
Compare
Product Number
Description
Pricing