Skip to Content
Merck
Sodium bisulfite solution purum, ~40%

Sodium bisulfite solution

Sodium bisulfite solution purum, ~40%
Synonym(s):
Sodium hydrogen sulfite
Linear Formula:
NaHSO3
CAS Number:
Molecular Weight:
104.06

Products

Product Category
Brand
Formula Weight
Physical Form
Quality Segment
Special Grade
Showing 1-2 of 2