Skip to Content
ES
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
mPAGE
®
4-12% Bis-Tris Precast Gel
Products
Product Category
precast gels (3)
Brand
Millipore (3)
Length (cm)
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
MP41G15
mPAGE
®
4-12% Bis-Tris Precast Gel,
L × W × wells 10 cm × 8 cm × 15 well
Expand
View Pricing
MP41G10
mPAGE
®
4-12% Bis-Tris Precast Gel,
L × W × wells 10 cm × 8 cm × 10 well
Expand
View Pricing
MP41G12
mPAGE
®
4-12% Bis-Tris Precast Gel,
L × W × wells 10 cm × 8 cm × 12 well
Expand
View Pricing