Skip to Content
ES
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Graphene quantum dots
CAS Number:
7440-44-0
EC Number:
231-153-3
Products
Product Category
quantum dots (4)
Brand
Sigma-Aldrich (4)
Color
Physical Form
Quality Segment
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
900726
Graphene quantum dots,
blue luminescent, powder
Expand
View Pricing
900560
Graphene quantum dots,
>0.005% in H
2
O
Expand
View Pricing
900712
Graphene quantum dots,
aqua green luminescent, 1 mg/mL in H
2
O
Expand
View Pricing
900713
Graphene quantum dots,
aqua green luminescent, powder
Expand
View Pricing