![Anti-ZBTB33 antibody produced in rabbit Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution](/deepweb/assets/sigmaaldrich/product/images/275/013/22fe53f8-78a5-40a4-ae6b-d4b529e719b5/640/22fe53f8-78a5-40a4-ae6b-d4b529e719b5.jpg)
Anti-ZBTB33 antibody produced in rabbit
![Anti-ZBTB33 antibody produced in rabbit Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution](/deepweb/assets/sigmaaldrich/product/images/275/013/22fe53f8-78a5-40a4-ae6b-d4b529e719b5/640/22fe53f8-78a5-40a4-ae6b-d4b529e719b5.jpg)
Synonym(s):
Anti-Transcriptional regulator Kaiso antibody produced in rabbit, Anti-Zinc finger and BTB domain-containing protein 33 antibody produced in rabbit, Anti-Zinc finger and BTB domain containing 33
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing