Skip to Content
ES
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Guanidine hydrochloride
Synonym(s):
Aminoformamidine hydrochloride, Aminomethanamidine hydrochloride, Guanidinium chloride, Guanidine hydrochloride
Linear Formula:
NH
2
C(=NH)NH
2
· HCl
CAS Number:
50-01-1
Molecular Weight:
95.53
EC Number:
200-002-3
Products
Product Category
detergents (3)
buffers (1)
salts (1)
Brand
Millipore (4)
SAFC (3)
Sigma-Aldrich (6)
Biological Source
Color
Feature
Formula Weight
Gene Alias
Melting Point (°C)
pH Value
Physical Form
Product Line
Purity
Quality Segment
Technique
Filter Products
Showing 1-13 of 13
Compare
Product Number
Description
Pricing
G3272
Guanidine hydrochloride,
for molecular biology, ≥99%
Expand
View Pricing
G4505
Guanidine hydrochloride,
≥99% (titration), organic base and chaeotropic agent
Expand
View Pricing
50950
Guanidine hydrochloride,
≥98%
Expand
View Pricing
1.37037
Guanidine hydrochloride,
EMPROVE
®
EXPERT, NF
Expand
View Pricing
PHG0006
Guanidine hydrochloride,
Expand
View Pricing
50933
Guanidine hydrochloride,
BioUltra, for molecular biology, ≥99.5% (AT)
Expand
View Pricing
50940
Guanidine hydrochloride,
≥99.0% (AT)
Expand
View Pricing
137272
Guanidine hydrochloride,
EMPROVE
®
EVOLVE
Expand
View Pricing
1.04220
Guanidine hydrochloride,
LAB
Expand
View Pricing
5010-OP
Guanidine hydrochloride,
OmniPur
®
Grade, >= 99.5%, Denaturant used for purification of nucleic acids and polypeptides from cell lysates
Expand
View Pricing
369080
Guanidine hydrochloride,
Powerful protein denaturant.
Expand
View Pricing
369075
Guanidine hydrochloride,
ULTROL
®
Grade, ≥99% (titration), protein denaturant for purification of proteins, nucleic acids, and mRNA
Expand
View Pricing
RDD001
Guanidine hydrochloride,
anhydrous, free-flowing, Redi-Dri
™
, ≥99%
Expand
View Pricing