Antipain dihydrochloride from microbial source
Synonym(s):
N-(Nα-Carbonyl-Arg-Val-Arg-al)-Phe, Antipain, Hydrochloride, Antipain dihydrochloride from microbial source, Antipain, Dihydrochloride
Empirical Formula (Hill Notation):
C27H44N10O6 · 2HCl
CAS Number:
Molecular Weight:
677.62
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Manufacturer
Physical Form
Research Area
Quality Segment
Mode of Action
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing