![5-[(2R)-2-Aminopropyl]-2-methoxybenzenesulfonamide certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/images/225/052/df6ae0c2-6c64-4358-b54b-fb999c0b47b7/640/df6ae0c2-6c64-4358-b54b-fb999c0b47b7.jpg)
5-[(2R)-2-Aminopropyl]-2-methoxybenzenesulfonamide
![5-[(2R)-2-Aminopropyl]-2-methoxybenzenesulfonamide certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/images/225/052/df6ae0c2-6c64-4358-b54b-fb999c0b47b7/640/df6ae0c2-6c64-4358-b54b-fb999c0b47b7.jpg)
Empirical Formula (Hill Notation):
C10H16N2O3S
CAS Number:
Molecular Weight:
244.31
Products
Brand
Formula Weight
Manufacturer
Product Line
Quality Segment
Application
Special Grade
Showing 1-1 of 1