Skip to Content
ES
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Eucalyptol
Synonym(s):
1,3,3-Trimethyl-2-oxabicyclo[2.2.2]octane, 1,8-Cineole, 1,8-Epoxy-
p
-menthane
Empirical Formula (Hill Notation):
C
10
H
18
O
CAS Number:
470-82-6
Molecular Weight:
154.25
EC Number:
207-431-5
Products
Product Category
building blocks (1)
lipids (1)
terpenes (1)
Brand
Sigma-Aldrich (3)
Boiling Point (°C)
Feature
Formula Weight
Melting Point (°C)
Physical Form
Purity
Agency
Quality Segment
Markush Class
Markush Group
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
C80601
Eucalyptol,
99%
Expand
View Pricing
8.43788
Eucalyptol,
for synthesis
Expand
View Pricing
53801
Eucalyptol,
tested according to Ph. Eur.
Expand
View Pricing