L-Cysteine
Synonym(s):
(R)-2-Amino-3-mercaptopropionic acid, L-Cysteine, L-Cysteine, Free Base, (R)-(+)-Cysteine
Linear Formula:
HSCH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
121.16
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Reaction Type
Special Grade
Technique
Showing 1-10 of 10
Compare
Product Number
Description
Pricing