U108
S(−)-UH-301 hydrochloride
solid
Synonym(s):
S(−)-5-Fluoro-8-hydroxy-2-dipropylamino-1,2,3,4-tetrahydronaphthalene hydrochloride, S(−)-5-Fluoro-8-hydroxy-DPAT hydrochloride
About This Item
Recommended Products
Assay
≥98% (HPLC)
form
solid
color
white
solubility
H2O: >20 mg/mL
ethanol: soluble
SMILES string
Cl[H].CCCN(CCC)[C@H]1CCc2c(F)ccc(O)c2C1
InChI
1S/C16H24FNO.ClH/c1-3-9-18(10-4-2)12-5-6-13-14(11-12)16(19)8-7-15(13)17;/h7-8,12,19H,3-6,9-11H2,1-2H3;1H/t12-;/m0./s1
InChI key
FKUVCCNCUAFKAH-YDALLXLXSA-N
Gene Information
human ... HTR1A(3350)
Biochem/physiol Actions
Caution
Preparation Note
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service