1000NN6
Conjugated Polymer Nanoparticle (CPN)
fluorescence λem 1000 nm (IR-II), carboxy-NHS-amine functionalized, 1 ea of 18 nmol oligonucleotide kit
Synonym(s):
CPN™ 30-minute conjugation kit
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
packaging
1 ea of 18 nmol oligonucleotide kit
fluorescence
λem 1000 nm (IR-II)
storage temp.
−20°C
Looking for similar products? Visit Product Comparison Guide
General description
CPNs are extremely bright and stable fluorophore nanoparticles which have been shown to give exceptional performance in a wide range of fluorescent analysis applications, such as lateral flow tests, fluorescent ELISA, cell tracking, immunofluorescence microscopy, flow cytometry and western blot. CPNs are resistant to photobleaching and a wide range of temperatures (-20°C to 120°C) and pH (2 to 12). They have been shown to be 1000x brighter than other fluorophores bringing exceptional levels of detection to assays and diagnostics.
Application
Amine Oligonucleotide Conjugation kit allows covalent linkage reactions to be readily carried out without access to specialist chemistry capabilities. The kit is designed to be specific for CPN – oligonucleotide linkage, has a rapid reaction time of 30 minutes and comes in three sizes for the conjugation of ~6nmole, ~18nmole and ~60nmole of oligonucleotide. CPNsTM are available in a range of emission wavelengths covering the visible and IR spectrum.
Legal Information
CPN is a trademark of Stream Bio Ltd
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service