Merck
  • Ferroportin mRNA is down-regulated in granulosa and cervical cells from infertile women.
Ferroportin mRNA is down-regulated in granulosa and cervical cells from infertile women.
Fertility and sterility (2016-11-16)
José Maria Moreno-Navarrete, Eva López-Navarro, Luz Candenas, Francisco Pinto, Francisco J Ortega, Mònica Sabater-Masdeu, Manuel Fernández-Sánchez, Victor Blasco, Antonio Romero-Ruiz, Marina Fontán, Wifredo Ricart, Manuel Tena-Sempere, José M Fernández-Real
ABSTRACT
To explore the relationship between iron and infertility by investigating iron-related gene expression in granulosa and uterine cervical cells. Case-control study. Two tertiary hospitals. Two independent cohorts of fertile (n = 18 and n = 17) and infertile (n = 31 and n = 35) women. In vitro fertilization. Gene expression levels of ferritin light chain (FTL), ferritin heavy chain (FTH), transferrin receptor (TFRC), and ferroportin (SLC40A1) mRNA were analyzed in granulosa and cervical cells. In the first cohort, fertile and infertile women were similar in body mass index. Ferroportin mRNA levels were decreased in granulosa cells from infertile women in parallel with increased serum hepcidin levels. A positive association between ferroportin and TFRC mRNA, a gene associated with intracellular iron deficiency, was observed only in granulosa cells from fertile women. The major findings were replicated in a second independent cohort. Ferroportin mRNAs and circulating hepcidin identify a subset of infertile women and may constitute a target for therapy.