Merck
  • A novel interaction between megakaryocytes and activated fibrocytes increases TGF-β bioavailability in the Gata1(low) mouse model of myelofibrosis.
A novel interaction between megakaryocytes and activated fibrocytes increases TGF-β bioavailability in the Gata1(low) mouse model of myelofibrosis.
American journal of blood research (2015-01-01)
Maria Zingariello, Alessandra Ruggeri, Fabrizio Martelli, Manuela Marra, Laura Sancillo, Ilaria Ceglia, Rosa Alba Rana, Anna Rita Migliaccio
ABSTRACT
Despite numerous circumstantial evidences, the pathogenic role of TGF-β in primary myelofibrosis (PMF), the most severe of the Philadelphia-negative myeloproliferative neoplasms, is still unclear because of the modest (2-fold) increases in its plasma levels observed in PMF patients and in the Gata1(low) mouse model. Whether myelofibrosis is associated with increased bioavailability of TGF-β bound to fibrotic fibres is unknown. Transmission electron-microscopy (TEM) observations identified that spleen from PMF patients and Gata1(low) mice contained megakaryocytes with abnormally high levels of TGF-β and collagen fibres embedded in their cytoplasm. Additional immuno-TEM observations of spleen from Gata1(low) mice revealed the presence of numerous activated fibrocytes establishing with their protrusions a novel cellular interaction, defined as peripolesis, with megakaryocytes. These protrusions infiltrated the megakaryocyte cytoplasm releasing collagen that was eventually detected in its mature polymerized form. Megakaryocytes, engulfed with mature collagen fibres, acquired the morphology of para-apoptotic cells and, in the most advanced cases, were recognized as polylobated heterochromatic nuclei surrounded by collagen fibres strictly associated with TGF-β. These areas contained concentrations of TGF-β-gold particles ~1000-fold greater than normal and numerous myofibroblasts, an indication that TGF-β was bioactive. Loss-of-function studies indicated that peripolesis between megakaryocytes and fibrocytes required both TGF-β, possibly for inducing fibrocyte activation, and P-selectin, possibly for mediating interaction between the two cell types. Loss-of-function of TGF-β and P-selectin also prevented fibrosis. These observations identify that myelofibrosis is associated with pathological increases of TGF-β bioavailability and suggest a novel megakaryocyte-mediated mechanism that may increase TGF-β bioavailability in chronic inflammation.
Sigma-Aldrich
Anti-Mouse Collagen Type I Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Monoclonal Anti-Fibronectin antibody produced in mouse, clone FN-15, ascites fluid