Merck
  • An individual patient data meta-analysis of adjuvant therapy with carmofur in patients with curatively resected colon cancer.
An individual patient data meta-analysis of adjuvant therapy with carmofur in patients with curatively resected colon cancer.
Japanese journal of clinical oncology (2005-09-13)
Junichi Sakamoto, Chikuma Hamada, Mahbubur Rahman, Susumu Kodaira, Katsuki Ito, Hiroaki Nakazato, Yasuo Ohashi, Masayuki Yasutomi
ABSTRACT
Oral carmofur, either as a single or in combination with other chemotherapeutic agents, has been used as adjuvant chemotherapy for curatively resected colon cancer patients. Past trials and meta-analyses indicate that it is somewhat effective in extending survival of patients with this cancer. The objective of this study was to perform a reappraisal of randomized clinical trials conducted in this regard. We designed an individual patient-based meta-analysis of relevant clinical trials to examine the benefit of oral carmofur for curatively resected colon cancer in terms of overall survival (OS) and disease-free survival (DFS). We analyzed individual patient data of three randomized clinical trials, which met the predetermined inclusion criteria. These three trials had a combined total of 2152 patients, carmofur as adjuvant chemotherapy compared with surgery-alone, 5 years follow-up, intention-to-treat-based analytic strategy and similar end points (OS and DFS). In a pooled analysis, 5 year OS rates were 80.4 and 76.4%, and 5 year DFS rates 76.9 and 71.0%, respectively, in carmofur and surgery-alone group. Oral carmofur had significant advantage over surgery-alone in terms of both OS [pooled hazard ratio, 0.82; 95% confidence interval (CI) = 0.68-0.99; P = 0.043] and DFS (pooled hazard ratio, 0.77; 95% CI = 0.65-0.91; P = 0.003). This individual patient-based meta-analysis demonstrated that oral carmofur significantly improves both OS and DFS in patients with curatively resected colon cancers.
Sigma-Aldrich
Carmofur, ≥98% (HPLC), powder