Merck
  • Biodegradability of end-groups of the biocide polyhexamethylene biguanide (PHMB) assessed using model compounds.
Biodegradability of end-groups of the biocide polyhexamethylene biguanide (PHMB) assessed using model compounds.
Journal of industrial microbiology & biotechnology (2006-05-10)
Leon P O'Malley, Andrew N Collins, Graham F White
ABSTRACT
Polyhexamethylene biguanide (PHMB), a biocide used in a wide variety of disinfection and preservation applications, is a polydisperse mixture in which the end-groups may be any combination of amine, guanidine and cyanoguanidine. Using PHMB model compounds (1,6-diaminohexane; 1,6-diguanidinohexane; 1,6-di(cyanoguanidino)hexane; 4-guanidinobutyric acid), we have determined the biodegradation characteristics of each end-group in several strains of bacteria isolated for their ability to utilise PHMB as a sole source of nitrogen. Bacteria were screened for growth at the expense of each model compound (at non-inhibitory concentrations) as sole nitrogen source. None of the isolated bacteria was capable of utilising a cyanoguanidine end-group as growth substrate, whereas several bacteria were shown to utilise amine or guanidine end-groups. In particular, a strain of Pseudomonas putida was capable of extensive growth with 1,6-diguanidinohexane as a sole nitrogen source, with complete removal of guanidine groups from culture medium within 2 days, and with concomitant formation of unsubstituted urea, which in turn was also utilised by the organism. We conclude that whilst amine and guanidine end-groups in PHMB are likely to be susceptible to biodegradation, cyanoguanidine end-groups are likely to be recalcitrant.
Sigma-Aldrich
4-Guanidinobutyric acid, ≥98%