Merck
  • Evaluation of phototoxic and cytotoxic potential of TiO2 nanosheets in a 3D reconstructed human skin model.
Evaluation of phototoxic and cytotoxic potential of TiO2 nanosheets in a 3D reconstructed human skin model.
ALTEX (2020-03-01)
Alžbeta Líšková, Silvia Letašiová, Soňa Jantová, Vlasta Brezová, Helena Kandarova
ABSTRACT
Despite a continuous increase of commercial products containing nanoparticles, only few materials are currently used in such large amounts, forms and sizes as titanium dioxide - TiO2. Besides its use in cosmetics, food industry and biomedicine, TiO2 nanoparticles (NPs) are also used as highly efficient photocatalysts due to their unique ability to convert complex organic materials to carbon dioxide, water and simple mineral acids via complex radical and electron transfer reactions. The main objective of the current project was to assess the potential dermal effect (phototoxicity and skin toxicity) of TiO2 nanosheets (TIG-800) synthesized from the lyophilized aqueous colloids of peroxo-titanic acid by high-temperature treatment. The effect was examined on reconstructed human skin model EpiDerm in the test pre-validated by ECVAM and adopted into the ICH S10 guidelines for the preclinical photo-safety assessment of drugs. In addition, two further commercial samples of nanocrystalline TiO2, namely Aeroxide P25 and Eusolex T-2000, and six benchmark materials from pre-validation studies were tested. None of the TiO2 NPs tested in the study caused acute phototoxicity or cytotoxicity in the reconstructed 3D tissues up to the highest concentration tested. The prediction of photo-irritation potency for the benchmark chemicals was comparable to previous studies. For some of the tested materials, we identified reasons for false negative results or variability in previously published datasets by improved dosing, conditions of irradiation, and choice of suitable solvents. The method proved its suitability for photo-irritation assessment of topically applied materials.