Merck
  • REG4 contributes to the invasiveness of pancreatic cancer by upregulating MMP-7 and MMP-9.
REG4 contributes to the invasiveness of pancreatic cancer by upregulating MMP-7 and MMP-9.
Cancer science (2012-09-11)
Xu-Jun He, Xiao-Ting Jiang, Ying-Yu Ma, Ying-Jie Xia, Hui-Ju Wang, Tian-Pei Guan, Qin-Shu Shao, Hou-Quan Tao
ABSTRACT
Recent studies have shown that overexpression of regenerating gene family member 4 (REG4) is associated with the initiation and progression of pancreatic cancer. In our study, we explored the role of REG4 in the invasion of pancreatic cancer. Real-time PCR and Western blot analysis were used to determine REG4 expression in pancreatic cancer cell lines. An MTT assay was carried out to test the effect of REG4 on the growth of pancreatic cancer cells. The involvement of REG4 in cancer cell invasion was examined by Transwell invasion assay. Two MMPs, MMP-7 and MMP-9, were identified from a pool of candidate genes as being related to REG4-induced cell invasion by PCR and Western blotting. Immunohistochemistry was used to confirm the correlation between REG4 and the two MMPs. High expression of REG4 was found in BXPC-3 cells and its culture media. But in PANC-1 and ASPC-1 cell lines, REG4 expression levels were very low, and no detectable protein was found in the culture medium. The MTT and Transwell invasion assays showed that recombinant REG4 protein and BXPC-3 conditioned media significantly promoted the proliferation and invasiveness of pancreatic cancer cells. It was also shown that MMP-7 and MMP-9 are upregulated by REG4 induction using real-time PCR and Western blotting analysis. Immunohistochemical study further verified this result. In conclusion, REG4 promotes not only growth but also in vitro invasiveness of pancreatic cancer cells by upregulating MMP-7 and MMP-9.
Sigma-Aldrich
QCM ECMatrix Cell Invasion Assay, 24-well (8 µm), fluorimetric, The Cell Invasion Assay Kit kuses a 24 well plate with 8 um pores, which is ideal for evaluation of invasive tumor cells.