Merck
  • RADAR-seq: A RAre DAmage and Repair sequencing method for detecting DNA damage on a genome-wide scale.
RADAR-seq: A RAre DAmage and Repair sequencing method for detecting DNA damage on a genome-wide scale.
DNA repair (2019-06-28)
Kelly M Zatopek, Vladimir Potapov, Lisa L Maduzia, Ece Alpaslan, Lixin Chen, Thomas C Evans, Jennifer L Ong, Laurence M Ettwiller, Andrew F Gardner
ABSTRACT
RAre DAmage and Repair sequencing (RADAR-seq) is a highly adaptable sequencing method that enables the identification and detection of rare DNA damage events for a wide variety of DNA lesions at single-molecule resolution on a genome-wide scale. In RADAR-seq, DNA lesions are replaced with a patch of modified bases that can be directly detected by Pacific Biosciences Single Molecule Real-Time (SMRT) sequencing. RADAR-seq enables dynamic detection over a wide range of DNA damage frequencies, including low physiological levels. Furthermore, without the need for DNA amplification and enrichment steps, RADAR-seq provides sequencing coverage of damaged and undamaged DNA across an entire genome. Here, we use RADAR-seq to measure the frequency and map the location of ribonucleotides in wild-type and RNaseH2-deficient E. coli and Thermococcus kodakarensis strains. Additionally, by tracking ribonucleotides incorporated during in vivo lagging strand DNA synthesis, we determined the replication initiation point in E. coli, and its relation to the origin of replication (oriC). RADAR-seq was also used to map cyclobutane pyrimidine dimers (CPDs) in Escherichia coli (E. coli) genomic DNA exposed to UV-radiation. On a broader scale, RADAR-seq can be applied to understand formation and repair of DNA damage, the correlation between DNA damage and disease initiation and progression, and complex biological pathways, including DNA replication.
Sigma-Aldrich
N6-Methyl-2′-deoxyadenosine