TSKgel® SuperSW3000 HPLC Column
Synonym(s):
TSKgel® Size Exclusion (SW-Type) HPLC Column
Products
Product Category
Manufacturer
Product Line
Agency
USP Suitability
Separation Technique
ID (mm)
Length (cm)
Particle Size (µm)
Pore Size (Å)
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing