Ubidecarenone
Synonym(s):
Coenzyme Q10, Q-10, Ubiquinone 50, Ubiquinone-10
Empirical Formula (Hill Notation):
C59H90O4
CAS Number:
Molecular Weight:
863.34
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Manufacturer
Physical Form
Purity
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-7 of 7
Compare
Product Number
Description
Pricing