Skip to Content
DK
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Safety Data Sheets (SDS)
Certificates (COA/COO)
Quality & Regulatory
Calculators & Apps
Webinars
Back
High Density Polyethylene
Synonym(s):
Polyethylene
Linear Formula:
H(CH
2
CH
2
)
n
H
CAS Number:
9002-88-4
Products
Brand
Supelco (1)
Manufacturer
Agency
Format
Quality Segment
Application
Special Grade
certified reference material (3)
analytical standard (1)
pharmaceutical primary standard (1)
Technique
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
1546707
High Density Polyethylene,
United States Pharmacopeia (USP) Reference Standard
Expand
View Pricing
ERMEC590
High Density Polyethylene,
ERM
®
, certified reference material
Expand
View Pricing
81223
High Density Polyethylene,
analytical standard, for GPC, 2,000
Expand
View Pricing
NIST1474B
High Density Polyethylene,
NIST
®
SRM
®
Expand
View Pricing
NIST1473C
High Density Polyethylene,
NIST
®
SRM
®
Expand
View Pricing