Skip to Content
Merck
Lysozyme from chicken egg white VETRANAL®, analytical standard

Lysozyme from chicken egg white

Lysozyme from chicken egg white VETRANAL®, analytical standard
Synonym(s):
Mucopeptide N-acetylmuramoylhydrolase, Muramidase
CAS Number:
EC Number:

Products

Product Category
Brand
Product Line
Format
Analyte Suitability
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing