Skip to Content
Merck
Benzoic acid phyproof® Reference Substance

Benzoic acid

Benzoic acid phyproof® Reference Substance
Synonym(s):
Benzoic acid
Linear Formula:
C6H5COOH
CAS Number:
Molecular Weight:
122.12

Products

Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Technique