Bruker standard solvent
Synonym(s):
Trifluoroacetic acid solution, TFA
Linear Formula:
CF3COOH
CAS Number:
Molecular Weight:
114.02
Products
Product Category
Brand
Formula Weight
Physical Form
Product Line
Chemical Composition
Quality Segment
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing