Deviron® C16 Detergent EMPROVE® EVOLVE solution
Synonym(s):
N,N-Dimethylmyristylamine N-oxide
Linear Formula:
CH3(CH2)13N(O)(CH3)2
CAS Number:
Molecular Weight:
257.46
Products
Product Category
Brand
Sustainability
Feature
Formula Weight
Physical Form
Product Line
Quality Segment
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing