![Chlorodipyrrolidinocarbenium hexafluorophosphate ≥97.5% (CHN)](/deepweb/assets/sigmaaldrich/product/structures/358/172/d06fc343-b7e8-4871-8263-3f50f4d84a31/640/d06fc343-b7e8-4871-8263-3f50f4d84a31.png)
Chlorodipyrrolidinocarbenium hexafluorophosphate
![Chlorodipyrrolidinocarbenium hexafluorophosphate ≥97.5% (CHN)](/deepweb/assets/sigmaaldrich/product/structures/358/172/d06fc343-b7e8-4871-8263-3f50f4d84a31/640/d06fc343-b7e8-4871-8263-3f50f4d84a31.png)
Synonym(s):
N,N,N′,N′-Bis(tetramethylene)chloroformamidinium hexafluorophosphate, Chloro-N,N,N′,N′-bis(tetramethylene)formamidinium hexafluorophosphate, PyClU
Linear Formula:
C9H16N2Cl · PF6
CAS Number:
Molecular Weight:
332.65
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Functional Group
Application
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing