Doxycycline hyclate
Synonym(s):
Doxycycline hydrochloride hemiethanolate hemihydrate, Doxycycline hyclate
Empirical Formula (Hill Notation):
C22H24N2O8 · HCl · 0.5H2O · 0.5C2H6O
CAS Number:
Molecular Weight:
512.94
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Manufacturer
Physical Form
Product Line
Activity Spectrum
Agency
Format
Quality Segment
Application
Mode of Action
API Family
Reference Material Type
Special Grade
Technique
Showing 1-8 of 8
Compare
Product Number
Description
Pricing