Skip to Content
Merck
Glyburide meets USP testing specifications

Glybenclamide

Glyburide meets USP testing specifications
Synonym(s):
5-Chloro-N-[4-(cyclohexylureidosulfonyl)phenethyl]-2-methoxybenzamide, Glyburide, N-p-[2-(5-Chloro-2-methoxybenzamido)ethyl]benzenesulfonyl-N′-cyclohexylurea, Glybenclamide
Empirical Formula (Hill Notation):
C23H28ClN3O5S
CAS Number:
Molecular Weight:
494.00
EC Number:

Products

Product Category
Brand
Color
Feature
Formula Weight
Gene Alias
Physical Form
Purity
Agency
Research Area
Quality Segment
Application